VWR® Real-Time PCR Plates, 96-Well and 384-Well
Supplier: VWR International
82006-650 82006-664 82006-678 82006-704 82006-708 82006-710 82006-712 82006-714 82006-718 83009-672 83009-676 93001-100 89218-288 89218-292 89218-294 89218-296 89218-298 10755-256 76402-858
82006-650CS 1123.72 CAD
82006-650 82006-664 82006-678 82006-704 82006-708 82006-710 82006-712 82006-714 82006-718 83009-672 83009-676 93001-100 89218-288 89218-292 89218-294 89218-296 89218-298 10755-256 76402-858
VWR® Real-Time PCR Plates, 96-Well and 384-Well
Microplates PCR Plates
VWR® 96- and 384-Well Real-Time PCR Plates are compatible with most leading real-time PCR instruments including most BioRad, Applied Biosystems, Roche Lightcycler, and Analytik Jena models.
  • Ultra-thin walls to optimize thermal transfer resulting in increased reaction efficiencies
  • Available with Ultra-clear wells for easy visualization of small sample volumes
  • Available in white options which increase the intensity of fluorescence transmitted through the cap or sealing film
  • Made of virgin polypropylene which is naturally low-binding without the need to incorporate additives
  • Black alphanumerics are easy-to-read and simplifies sample identification
  • All plates are certified free of RNAse, DNAse, and human DNA
Compatible with VWR® 8-strip flat caps (83009-684).