InocuLoop Inoculating Loops, Caplugs
Supplier: Caplugs
333-5001-B10 333-5002-B1I 333-5010-Y10 333-5011-Y1I 333-5012-G10 333-5013-G1I
76492-636CT 145.68 CAD
76492-636 76492-638 76492-640 76492-642 76492-644 76492-646
InocuLoop Inoculating Loops, Caplugs
Spreaders Inoculating Loops and Needles
Inoculoop disposable inoculating loops have patented four-sided spreaders, enabling the inoculation of a petri dish with one, not multiple, disposable loops. Make bacterial inoculation and quantitative procedures faster and more efficient.
  • Made of flexible plastic for easy spreading
  • Can also be used as a picker for isolating colonies
  • Pre-sterilized Inoculoops are gamma-irritated
Suitable in anaerobic chambers, in the field and even in satellite metal loops.
Order Now
SPECIFICATIONS
Environmentally Preferable