Immunostain Moisture Chambers, Caplugs
Supplier: Caplugs
240-9000-010 240-9020-Z10 240-9035-N10
76492-648CT 347.41 CAD
76492-648 76492-650 76492-652
Immunostain Moisture Chambers, Caplugs
Staining Boxes
Moisture chambers are designed to maintain moisture levels for immunostaining and diagnostic reaction processes.
  • Chamber is divided into ten (10) individual compartments with approximately 0.5" space between compartments
  • Chamber is fabricated from heavy-duty polystyrene with an air-tight design to keep moisture in
  • Chambers are stackable in order to save counter and/or refrigerator space
  • Available in clear, black, and amber
When the chamber lid is closed, eight barrier dividers are placed into the empty spaces between slides, completely isolating allMore Product Information