Brady® Thermal Transfer 1 Core R6000 Halogen-Free Series Printer Ribbon
Supplier: Brady
R6007 55752 R6602
76460-980RL 413.76 CAD
76460-980 76243-618 76460-978
Brady® Thermal Transfer 1 Core R6000 Halogen-Free Series Printer Ribbon
Printer Ribbons
This ink-side-out ribbon features an open 1" core for compatibility with a wide variety of thermal transfer printers.
  • Halogen-free
  • Heat-resistant
  • Scratch-resistant
  • Solvent-resistant
One-color ribbon for printing a single color onto a label. R6000 Series Halogen-Free Resin Formulation offers excellent solvent, scratch and heat resistance when printed on the appropriate label material. Recommended for printing on glossy or very smooth label stocks, such as B-483 Ultra aggressive polyester and B-423 Permanent polyester from Brady®. Combine Brady label materials with their recommended ribbon series for superior legend performance.
These ribbons are designed for use with BBP®81 Label Printer, BradyPrinter i7100 Industrial Label Printer, and THT 3" Core Printers.