Heat Shrink Polyolefin Wire Marking Sleeves, Jumbo Roll, Brady
Supplier: Brady Worldwide
SA-3PS-250-1-YL-J SA-3PS-375-1-YL-J
76461-352RL 8586.27 CAD
76461-352 76461-354
Heat Shrink Polyolefin Wire Marking Sleeves, Jumbo Roll, Brady
Labels
PermaSleeve® Heat Shrink Tubing is engineered for thermal transfer or dot matrix printing and is ideal for use in wire identification.
  • Flame-retardant
  • Heat-shrinkable
  • Wire diameter range: 0.094 to 0.215"
Sleeves are designed for use with BradyPrinter i7100 Industrial Label Printer, BSP45 Wire and Cable Sleeve Applicator, and THT 3" Core Printers.
Ordering information: A ribbon is required to print this material. Use Part Number IP-R6606 for the BradyPrinter i5100 or BRADY IP® Series Label Printer; use R6606 for a different printer.