High Adhesion Glossy Polyester with Rubber Adhesive Labels, 3" Core, Brady
Supplier: Brady Worldwide
THT-5-483-10 THT-19-483-1 THT-55-483-1 THT-76-483-1
76461-080RL 424.33 CAD
76461-080 76461-086 76461-088 76461-090
High Adhesion Glossy Polyester with Rubber Adhesive Labels, 3" Core, Brady
Labels
Ultra Aggressive Polyester (B-483) is perfect for labeling products and general identification, and can be applied to powder-coated, smooth, textured, or irregular surfaces.
  • Aggressive adhesive
Ultra Aggressive Polyester (B-483) is a glossy white polyester with permanent rubber based adhesive. The labels are thermal transfer printable. Material is designed for high adhesion to textured metals and low surface energy plastics. Service temperature of –40 to +248 °F (–40 to +120 °C).
Labels are designed for use with BBP®11 Label Printer, BBP®12 Label Printer, BBP®81 Label Printer, Brady IP® Series Label Printer, BradyPrinter i5100 Industrial Label Printer, BradyPrinter i7100 Industrial Label Printer, and THT 3" Core Printers.
Certifications: UL approved. CSA accepted.
Ordering information: A ribbon is required to print this material. Use Part Number IP-R6007 for the BradyPrinter i5100 or BRADY IP® Series Label Printer; use R6007 for a different printer.