Raised Panel Polyester Rectangular Labels, 3" Core, Brady
Supplier: Brady Worldwide
THTEP-171-593-.5 THTEP-173-593-.5 THTEP-176-593-.5
76461-152RL 460.61 CAD
76461-152 76461-154 76461-158
Raised Panel Polyester Rectangular Labels, 3" Core, Brady
Labels
Raised panel Adhesive-Taped Polyester (B-593) labels are designed for patch panel identification, external push-buttons, switches, and internal connection points.
  • Raised panel design
  • Aggressive adhesive
  • Solvent-resistant
Raised panel Adhesive-Taped Polyester (B-593) labels can be used as rating and serial plates using alphanumerics that require nameplate quality. Aggressive adhesive and solvent resistance makes them a great replacement for engraved tags. Service temperature of ‒20 to +100 °C (‒4 to +212 °F)
Labels are designed for use with BBP®81 Label Printer, Brady IP® Series Label Printer, BradyPrinter i5100 Industrial Label Printer, BradyPrinter i7100 Industrial Label Printer, THT 3" Core Printers.
Certifications: UL approved.
Ordering information: A ribbon is required to print this material. Use Part Number 49006-110 for the BradyPrinter i5100 or BRADY IP® Series Label Printer; use R6006 for a different printer.