Raised Panel Polyester Rectangular Labels, 3" Core
Supplier: Brady Worldwide
THTEP-178-593-.5 THTEP-172-593-.5 115872
76461-148PK 1650.04 CAD
76461-148 76461-150 77532-228
Raised Panel Polyester Rectangular Labels, 3" Core
Labels
Raised panel adhesive-taped polyester (B-593) labels are designed for patch panel identification, external push-buttons, switches, and internal connection points.
  • Raised panel design
  • Aggressive adhesive
  • Solvent-resistant
Raised panel adhesive-taped polyester (B-593) labels can be used as rating and serial plates using alphanumerics that require nameplate quality. Aggressive adhesive and solvent resistance makes them a great replacement for engraved tags. Service temperature of ‒20 to +100 °C (‒4 to +212 °F).
Labels are designed for use with BBP®81More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable