High Adhesion Glossy Polyester with Rubber Adhesive Labels, 3" Core, Brady
Supplier: Brady Worldwide
THT-17-483-3 THT-18-483-3
76461-082RL 377.85 CAD
76461-082 76461-084
High Adhesion Glossy Polyester with Rubber Adhesive Labels, 3" Core, Brady
Labels
Ultra Aggressive Polyester (B-483) is perfect for labeling products and general identification, and can be applied to powder-coated, smooth, textured, or irregular surfaces.
  • Aggressive adhesive
Ultra Aggressive Polyester (B-483) is glossy white polyester with a permanent rubber based adhesive. The labels are thermal transfer printable. Material is designed for high adhesion to textured metals and low surface energy plastics. Service temperature of ‒40 to +120 °C (‒40 to +248 °F).
Labels are designed for use with BBP®11 Label Printer, BBP®12 Label Printer, BBP®81 Label Printer, Brady IP® Series Label Printer, BradyPrinter i5100 Industrial Label Printer, BradyPrinter i7100 Industrial Label Printer, and THT 3" Core Printers.
Certifications: UL approved. CSA accepted.
Ordering information: A ribbon is required to print this material. Use Part Number 89077-379 for the BradyPrinter i5100 or BRADY IP® Series Label Printer; use 101076-574 for a different printer.