Glossy White Polyester Barcode and Solar Panel Labels, 3" Core, Brady
Supplier: Brady Worldwide
THT-3-423-10 THT-19-423-1 THT-22-423-2 THT-55-423-1 THT-45-423-10 THT-53-423-3 THT-7-423-3 THT-1-423-10 THT-37-423-10 THT-17-423-3
76461-032RL 257.02 CAD
76461-032 76461-042 76461-046 76461-052 76461-064 76241-649 76241-659 76243-671 76243-673 76245-055
Glossy White Polyester Barcode and Solar Panel Labels, 3" Core, Brady
Labels
Permanent Polyester (B-423) is ideal for electronic PCB and component identification, bar code labels, rating plates, and solar panel identification.
  • Aggressive adhesive
  • Harsh environment use
  • Heat-resistant
  • Permanent adhesive
Glossy white finish with permanent acrylic is thermal transfer printable. Service temperature of ‒70 to +110 °C (‒94 to +230 °F).
Labels are designed for use with BBP®11 Label Printer, BBP®12 Label Printer, BBP®81 Label Printer, Brady IP® Series Label Printer, BradyPrinter i5100 Industrial Label Printer, BradyPrinter i7100More Product Information