Heatex Polyolefin 20 to 16 Gauge Wire Marking Sleeves, 3" Core, Brady
Supplier: Brady Worldwide
HX-2000-2-YL HX-125-150-WT HX-125-2-WT HX-187-2-WT-4 HX-187-2-WT-2 HX-187-150-WT HX-187-2-WT HX-250-2-WT-2 HX-250-150-WT HX-250-2-WT HX-375-2-WT-2 HX-375-150-WT HX-375-2-WT HX-500-150-WT 2HX-187-2-WT 2HX-375-2-WT HX-187-150-WT-J-2 HX-187-2-WT-J-2 HX-375-150-WT-J
76461-300RL 728.93 CAD
76461-300 76461-302 76461-304 76461-306 76461-308 76461-310 76461-312 76461-314 76461-316 76461-318 76461-320 76461-322 76461-324 76461-326 76461-328 76461-330 76461-332 76461-334 76461-336
Heatex Polyolefin 20 to 16 Gauge Wire Marking Sleeves, 3" Core, Brady
Labels
Heat-Shrink PermaSleeve® HX Polyolefin (B-7642) Wire Marking Sleeves are 2:1 heat shrinkable, high-temperature rated, highly flame retardant, and very flexible.
  • Heat-shrinkable
Heat-Shrink PermaSleeve® HX Polyolefin (B-7642) Wire Marking Sleeves are supplied in roll form in a flattened format on a carrier designed for use with computer driven printers. Operating temperature of ‒40 to +120 °C (‒40 to +248 °F).
Sleeves are designed for use with BBP®11 Label Printer, BBP®12 Label Printer, BBP®72 Label Printer, BradyPrinter i7100 Industrial LabelMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable