Matte Amber Polyimide Circuit Board Labels, 3" Core, Brady
Supplier: Brady Worldwide
THT-28-724-10 THT-4-724-10 THT-14-724-10
76461-114RL 2252.05 CAD
76461-114 76461-116 76241-633
Matte Amber Polyimide Circuit Board Labels, 3" Core, Brady
Labels
Amber high temperature polyimide with an ultra-durable adhesive withstands the extreme wash protocol and cleaning chemicals used in printed circuit board manufacturing.
  • Heat-resistant, permanent adhesive
Amber high temperature polyimide withstands multiple cycles of harsh condition washes for printed circuit boards and electronic component pre-process labeling. Resists high temperatures up to 300 °C (572 °F). Survives on the top or bottom side of printed circuit boards through the solder process.
Labels are designed for use with BBP®11 LabelMore Product Information