Metallized High Adhesion Matte Polyester Labels, 3" Core, Brady
Supplier: Brady Worldwide
THT-17-486-3 THT-18-486-3 THT-19-486-1
76461-024RL 591.57 CAD
76461-024 76461-026 76461-028
Metallized High Adhesion Matte Polyester Labels, 3" Core, Brady
Labels
Ultra-aggressive metallized polyester (B-486B) is ideal for rating/serial plates that use barcodes, alphanumerics, graphic symbols, and logos, and require nameplate-like quality.
  • Aggressive, heat-resistant, permanent adhesive
  • Service temperature: ‒40 to +120 °C (‒40 to +248 °F)
Matte, light gray material has a permanent rubber-based adhesive and is thermal transfer printable. Designed for high adhesion to textured metals, powder-coated surfaces, and low energy plastics.
Labels are designed for use with BBP®11 Label Printer, BBP®12 Label Printer, BBP®81 Label Printer, Brady IP® Series Label Printer, BradyPrinter i5100 Industrial Label Printer, BradyPrinter i7100 Industrial Label Printer, or THT 3" Core Printers.
Ordering information: A ribbon is required to print this material. Use Part Number 89077-374 for the BradyPrinter i5100 or BRADY IP® Series Label Printer; use 94000-380 for a different printer.