Self-Laminating Vinyl Wire and Cable Labels, 3" Core, Brady
Supplier: Brady
THT-65-427-3 THT-82-427-0.5 THT-66-427-1.5 THT-81-427-1 THT-64-427-3 THT-119-427-2.5 THT-56-427-10 30112 THT-74-427-10 THT-73-427-10 THT-80-427-5 THT-116-427-5 THT-113-427-5 THT-75-427-3 THT-8-427-10 THT-121-427-2.5
10796-511RL 505.37 CAD
10796-511 76461-070 76461-170 76461-060 76461-062 76461-078 76245-990 76461-034 76461-068 76461-058 76461-168 76461-074 76461-162 76461-076 76461-054 76461-066
Self-Laminating Vinyl Wire and Cable Labels, 3" Core, Brady
Labels
Self-Laminating Vinyl (B-427) is an excellent material for wire and cable identification. This material has good clarity, conformability, and is self-extinguishing.
  • Flame-retardant, self-laminating material
  • Service temperature: ‒40 to +70 °C (‒40 to +158 °F)
Provided in a self-laminated format with a white printable zone and translucent overlaminating area to protect text. Durable, permanent acrylic adhesive has outstanding water and oil resistance.
Labels are designed for use with BBP®11 Label Printer, BBP®12 Label Printer, BBP®81 Label Printer, Brady IP® Series Label Printer, BradyPrinter i5100 Industrial Label Printer, BradyPrinter i7100 Industrial Label Printer, or THT 3" Core Printers.
Ordering information: A ribbon is required to print this material. Use Part Number 89079-927 for the BradyPrinter i5100 or BRADY IP® Series Label Printer; use 49006-270 for a different printer.