DRNAse Free™ Surface Decontaminant, Cole Parmer
Supplier: Antyila Scientific
78901-43 78901-44
76489-338EA 82.04 CAD
76489-338 76489-340
DRNAse Free™ Surface Decontaminant, Cole Parmer
Cleaning Agents Decontaminants
Instantly eliminate RNase and DNase from labware and surfaces.
  • Ideal for items and surfaces used in PCR that cannot be autoclaved
  • Spray and rinse—works on glass, plastic, stainless steel, and benchtops
  • Replace carcinogenic chemicals, like DEPC, with this nontoxic formula
  • Quit baking glassware—doesn''t leave a residue so it won't interfere with gel polymerization or enzymatic reactions
This nontoxic formula doesn’t leave a residue, so it won’t interfere with gel polymerization or enzymatic reactions.