Reagent Reservoirs, Globe Scientific
Supplier: Globe Scientific
14010B 140101 140105 14025B 140251 140255 14050B 140501
76491-558BX 59.8 CAD
76491-558 76491-560 76491-562 76491-564 76491-566 76491-568 76491-570 76491-572 76491-574
Reagent Reservoirs, Globe Scientific
Reagent Reservoirs
These reagent reservoirs are suitable for virtually all of your lab’s sample preparation and aliquoting needs.
  • Pour spouts at every corner
  • Rigid sidewall design
  • Wide, stable base
  • Molded graduations
  • Compatible with multi-channel pipettors
  • Dnase, Rnase, purogen, animal materials and latex free
  • Produced from virgin medical grade polystyrene
Design enhancements include pour spouts at each corner, a rigid sidewall design and graduations for easy volume reference. Sterile variants are irradiated to guarantee a sterility assurance level (SAL) of 10-⁶.More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable