Corning® Permeable Supports, Polycarbonate (PC) Membrane, Corning
Supplier: Corning
3401 3402 3412 3413 3414 3415 3421 3428 3422 7910
29442-086CS 645.91 CAD
29442-086 29442-088 29442-104 29442-106 29442-108 29442-110 29442-118 29442-130 29442-120 76404-722
Corning® Permeable Supports, Polycarbonate (PC) Membrane, Corning
Microplates Insert Plates and Inserts
75 mm Transwell® with 0.4 µm pore polycarbonate membrane insert, sterile.
  • 10 µm thick translucent polycarbonate membrane
  • Treated for optimal cell attachment
  • Each insert is packaged in a 100 mm dish, 12 inserts per case
  • Membrane must be stained for cell visibility
Produces a cell culture environment that closely resembles an in vivo state. An essential tool for the study of both anchorage-dependent and independent cell lines.
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification RECs, ISO 14001 - Kennebunk