PT100 Sensors for External Control/Monitoring, JULABO
Supplier: JULABO
8891001 8891002 8891003 8891004 8891005 8891006 8891018 8891019 8891031 8891032
76480-274EA 1494.11 CAD
76480-274 76480-276 76480-278 76480-280 76480-282 76480-284 76480-286 76480-288 76480-290 76480-292
PT100 Sensors for External Control/Monitoring, JULABO
Temperature Probes
Temperature control across a broad temperature range.
  • Control and monitor temperature from your application
  • For use with JULABO products with PT100 port
Many of TCUs, have external PT100 sensor capabilities. JULABO offers PT100 sensors in stainless steel and PTFE-coated options. PTFE coatings are used for corrosive conditions. Sensors with 12’ shielded cable and 4-pin LEMOSA connector. These sensors ensure your application reaches the desired set point with a JULABO circulator for direct control and monitoring from the external application.
Order Now
SPECIFICATIONS
Environmentally Preferable