CrossLab Purged Packed Inlet for GC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
19243-80530 19243-80570 19244-80540 5080-8898 5080-8732 5181-3382
CA19243-80530EA 337.52 CAD
CA19243-80530 CA19243-80570 CA19244-80540 CA5080-8898 CA76514-710 CA76514-748
CrossLab Purged Packed Inlet for GC, Agilent Technologies
Chromatography Supplies Chromatography Spare Parts
Purged packed inlets are simple in both design and use.
  • Designed, tested and manufactured by Agilent engineers to work perfectly with our instruments superior performance
Packed column analysis is frequently done when high-efficiency separations are not needed or when gases are analyzed by gas-solid chromatography. Purged packed inlets for GC are simple in both design and use. Few parameters need to be set, and all carrier gas flow flushes through the inlet into the column in the standard configuration.