Transport Vial Caps, Caplugs
Supplier: Caplugs
300-3010-02N 300-3010-B2N 300-3010-E2N 300-3010-G2N 300-3010-L2N 300-3010-R2N 300-3010-W2N 300-3010-Y2N
76480-234BG 230.36 CAD
76480-234 76480-236 76480-238 76480-240 76480-242 76480-244 76480-246 76480-248
Transport Vial Caps, Caplugs
Closures Test/Sample Tube Closures
Caplugs Transport Vial Caps are designed to fit all sizes of transport vials.
  • Caps also fit 15 ml threaded centrifuge tubes
  • All transport vials are 95 kPa compliant
Available in a wide range of colors.