SP Bel-Art 96 Deep-Well Plates, Automation Compliant, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F37860-0007 F37860-0008
76487-080CS 836.61 CAD
76487-080 76487-082
SP Bel-Art 96 Deep-Well Plates, Automation Compliant, Bel-Art Products, a part of SP
Microplates Deep Well Microplates
Used with multi-channel pipettes or with automation systems, these 96 Deep-Well Plates are made of virgin polypropylene.
  • SBS/ANSI/SLAS format compatible
  • DNase, RNase, pyrogen free, manufactured in Class 8 clean room
  • Alphanumeric marking and cut-angle mark for easy referencing
  • Can be used with multi-channel pipettes or automation systems
  • Can be stored at –80 °C
  • Autoclavable up to 121 °C
V-bottom shape is ideal for ease of sample retrieval. U-bottom shape is ideal for agglutination or applications where light stirring is necessary.