CE & CE/MS Supplies, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
G1600-67201 1520-0401 G1607-20030 G1607-60041 G7100-68705
CAG1600-67201EA 371.03 CAD
CAG1600-67201 CAAG1520-0401 CAAGG160720030 CAAGG160760041 CAAGG710068705
CE & CE/MS Supplies, Agilent Technologies
Electrophoresis Systems Capillary Electrophoresis Systems
CE/MS sprayer kit and CE/MS adapter kit to couple an Agilent CE instrument with an MS system.
Agilent provides CE and CE/MS supplies, including sprayer kits and CE adapter kits, to couple an Agilent CE instrument with MS systems.The CE/MS sprayer kit needs the CE/MS adapter kit to fully support CE/MS coupling.