NEBNext® Ultra II RNA Library Prep with Sample Purification Beads, New England Biolabs
Supplier: New England Biolabs (NEB)
E7775L E7775S
103501-454EA 5672.2 CAD
103501-454 103501-456
NEBNext® Ultra II RNA Library Prep with Sample Purification Beads, New England Biolabs
Nucleic Acid Reagents Next Generation Sequencing Reagents
The Ultra II RNA Library Prep Kit for Illumina is designed for non-directional (non-strand-specific) RNA library construction, and delivers significantly increased sensitivity and specificity from your RNA-seq experiments, from ever-decreasing amounts of input RNA.
  • Generate the highest yields of high quality libraries, with a broad range of input amounts
  • Increase the complexity and transcript coverage of your libraries
  • Optimize your time with streamlined workflows, reduced hands-on time, and automation compatibility
  • Rely on robust performance, even with low quality RNA, including FFPE
  • Enjoy the flexibility and reliability of the gold standard SPRIselect beads
In conjunction with ribosomal RNA (rRNA) depletion or poly(A) mRNA enrichment, the kit enables the production of high quality libraries from 5 ng or 10 ng of Total RNA, respectively, up to 1 µg. For convenience, this kit includes SPRISelect beads.