NEBNext® Ultra II DNA Library Prep with Sample Purification Beads, New England Biolabs
Supplier: New England Biolabs (NEB)
The NEBNext Ultra II DNA Library Prep with Sample Purification Beads contains the enzymes, buffers and purification beads required to convert a broad range of DNA input amounts into indexed libraries for next-generation sequencing on the Illumina platform.
- Get more of what you need, with the highest library yields
- Use to generate high quality libraries from a broad range of input amounts, from 500 pg to 1 µg
- Prepare libraries from ALL your samples, including GC-rich DNA and FFPE DNA samples
- Improve yields and quality for target enrichment applications
- Includes SPRIselect beads
The fast, user-friendly workflow has minimal hands-on time and, for added convenience, this kit contains NEBNext Sample Purification Beads (SPRIselect beads from Beckman Coulter) for size selection and enzyme reaction cleanup.