Microcentrifuge Tube Racks
Supplier: MTC BIO, INC.
R1040 R1040-B R1040-O R1040-R R1040-G R1040-Y
470334-388PK 60.15 CAD
470334-388 470334-390 470334-392 470334-394 470334-396 470334-398
Microcentrifuge Tube Racks
Racks Tube Racks
Compact microcentrifuge tube racks to keep you and your samples organized.
  • Autoclavable
  • Accomodates conical and self-standing tubes
  • Raised, molded numbers
These compact, convenient 80 well racks are designed to keep your tubes neatly and efficiently organized. They are compatible with conical and self-standing microtubes and all wells have raised, molded numbers for quick identification.
The bottom of each well has been recessed to improve conical tube stability without sacrificing the stability of self-standing tubes. They are made of autoclavableMore Product Information