Diamond®MAX™ Centrifuge Tubes, Globe Scientific
Supplier: Globe Scientific
6295 6296 6297 6298 6299
76452-246CS 198.96 CAD
76452-246 76452-248 76452-250 76452-252 76452-254
Diamond®MAX™ Centrifuge Tubes, Globe Scientific
Tubes Centrifuge Tubes
Diamond® Max™ Centrifuge tubes are engineered to meet the demands of the most critical scientific work. Premium grade materials and an automated, touch free manufacturing process provide both performance and savings to users.
  • Ultra high performance: High spin rates with maximum RCF ratings of up to 20000 ×g
  • Premium materials: USP Class VI resin is also FDA CFR 21 compliant for food safety
  • Leak resistant: Easy Grip Caps have an integral seal ring design for secure closures that eliminate seepage when used as recommended
  • Value: Efficient automated production
More Product Information