Acetic acid ≥96%, EMPROVE® ESSENTIAL, SAFC®

Supplier: EMD MILLIPORE – PCS CA EXCEP
Danger

1.00058.2500 1.00058.9025
CA1.00058.2500CS 1913.22 CAD
CA1.00058.2500 CA1.00058.9025
Acetic acid ≥96%, EMPROVE® ESSENTIAL, SAFC®
Acetic acid
Formula: H₃CCOOH
Storage Temperature: Ambient
MDL Number: MFCD00036152
CAS Number: 64-19-7
EINECS: 200-580-7
UN: 2789
ADR: 8 (3),II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR