Autoclavable Safety Goggles, Cole-Parmer
Supplier: Antyila Scientific
16392-21 16392-22
76485-866BX 586.81 CAD
76485-866 76485-868
Autoclavable Safety Goggles, Cole-Parmer
Goggles
Prevent contamination entering the cleanroom environment.
  • Tested to withstand 35 autoclave cycles at 121 °C, for 30 minutes
  • No deformation or change in transparency due to autoclaving
  • Soft thermoplastic rubber frame provides utmost comfort
Cole-Parmer safety goggles provide optimal comfort while being durable and autoclavable. These wide-vision goggles provide a high level of comfort and maximum protection while being able to fit over most prescription glasses. Constructed of a thermoplastic rubber frame, scratch-resistant polycarbonate lens with premium anti-fog coating, and 24 mm non-irritant silicone strap.
Certifications: EN 166:2001 and ANSI Z87.1:2015.