J.T.Baker® Plate Square Well
Supplier: Ritter, part of Avantor
43001-00201 43001-00621 43001-00661
43001-0020CS 323.13 CAD
43001-0020 43001-0062 43001-0066
J.T.Baker® Plate Square Well
Microplates Deep Well Microplates
These 96 deep square well plates, made from virgin PP, are designed to make the interchanging of plates simpler in automated systems. They are also ideal for housing both cell and bacterial culture.
  • Chemical resistant
  • V-bottom well shape maximises liquid removal and aids resuspension
  • No inner edges to interfere with magnetic bead collection
  • Raised well rims to improve heat sealing
  • Conical base aids sample concentration, reconstitution and centrifugation
For a maximum capacity utilization.