J.T.Baker® Plate 96
Supplier: Ritter, part of Avantor
43001-06001 43001-01171 43001-01181 43001-01191
43001-0600CS 609.2 CAD
43001-0600 43001-0117 43001-0118 43001-0119
J.T.Baker® Plate 96
Microplates Deep Well Microplates
J.T.Baker® Plate 96 round well plates made from PP are ideal for ELISA applications.
  • Alphanumeric coding assists with identification of samples
  • Plate shape significantly reduces sample carryover
  • Standardised dimensions (128×85 mm) ensure compatibility with most common laboratory machines
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact Low Manufacturing Impact
Environmental certification ISO 50001 (Certificate Registration No.) 10000406726-MSC-RvA-DEU ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)