J.T.Baker® Robotic Tips Type HT
Supplier: Ritter, part of Avantor
49009-00051 49009-01051 49009-11051 49008-00031 49008-01051 49010-00031 49010-01051 49008-11051 49010-11051 49008-10051 49009-10051
49008-0104CS 1984.27 CAD
49008-0104 49008-1104 49009-0005 49009-0105 49009-1105 49010-0004 49010-0104 49010-1104 49008-0003 49008-0105 49010-0003 49010-0105 49008-1105 49010-1105 49008-1005 49009-1005
J.T.Baker® Robotic Tips Type HT
Pipette Tips
Suitable for use on third party robotic instruments Hamilton Microlab Star and Nimbus. Available in conductive version, with or without filter.
  • Manufactured under cleanroom conditions (cleanroom class 8)
  • State of the art quality control
  • Ideal for high-throughput analysis
  • Barcoded trays for automated identification
  • Packed in blister
J.T.Baker® robotic tips type HT are suitable for third party robotic instruments Hamilton Microlab Star, Nimbus, and other robotic instruments using Hamilton’s CO-RE pipetting head.
The validation of the tips on the instrument for specific applications is within the responsibility of the user.
Hamilton, Microlab Star™, Nimbus® and CO-RE® are registered trademarks of Hamilton company.