Motic Panthera E2 Upright Compound Microscopes
Supplier: Motic
1100104601116 76480-310 1100104601116 1100104601117 1100104601126 1100104601127
76480-304EA 2112.19 CAD
76480-304 76480-306 76480-308 76480-310 CA76480-306 CA76480-304 CA76480-310 CA76480-308
Motic Panthera E2 Upright Compound Microscopes
Microscopes
With the new microscope Series Panthera E2, your decision is distinct. This microscope series allows to experience Life Sciences in a new way, with best possible learning effect.
  • Reversed quadruple nosepiece
  • LED light indicator
  • CCIS Plan UC objectives 4×, 10×, 40× S
  • Coaxial coarse and fine focusing adjustment with tension adjustment
  • Built in low position coaxial rackless stage with right hand control
  • Focusable Abbe condenser N.A.0.90 with iris diaphragm and slot
Panthera E2 microscopes combine high quality materials and precise engineering to a persistent and resilient construction. Smooth operation of the mechanics is self-evident for fast and reliable results, responsible for shifting your daily work in classroom and lab to a higher level of quality. User-friendly design, robust for tough environments and unique features grants the best performance. Models of this series convince positively in smooth operation and haptic.
Ordering information: Contact your Avantor representative to learn more about these and other Motic solutions.