CompactDry™ Total Count (TC), Hardy Diagnostics
Supplier: Hardy Diagnostics
CompactDry™
TPC100 54081
CA10145-968PK 244.71 CAD
76414-994 CA10145-968
CompactDry™ Total Count (TC), Hardy Diagnostics
Microbiology Test Systems Microorganism Tests
CompactDry™ TC is a ready-to-use test method recommended for the determination of total aerobic bacterial counts.
  • No leakage
  • Easy to label
  • Self-diffusing media
  • Sturdy frame, stackable packaging
  • Ability to subculture without damage to colonies
Each cassette comes with 4 connected plates that can be easily snapped apart to support a variety of dilution scenarios.
Caution: Stored at room temperature vs the competitor products with storage at temperatures less than or equal to 8 °C.