Dichloroacetic acid (DCA) 3% in toluene deblocking reagent, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

BI0832-NP20 BI0832-2505
CABI0832-NP20EA 6136 CAD
CABI0832-NP20 CABI0832-2505 CABI0832-4000
Dichloroacetic acid (DCA) 3% in toluene deblocking reagent, Sigma-Aldrich®
Dichloroacetic acid (DCA)

These deblocking reagents are mainly involved in DNA synthesis.


  • Infrared spectrum conforms to the standard
  • Corrosive and toxic
  • Clear liquid form

Formula: C₂H₂Cl₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00004223
CAS Number: 79-43-6
UN: 2924
ADR: 3.8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR