Trichloroacetic acid (TCA) 3% in dichloromethane deblocking reagent

Supplier: MilliporeSigma

BI0830-0450 BI0830-0950 BI0830-4000 BI0830-P4000
CABI0830-0450CS 986 CAD
CABI0830-0450 CABI0830-0950 CABI0830-4000 CABI0830-P4000
Trichloroacetic acid (TCA) 3% in dichloromethane deblocking reagent
Trichloroacetic acid (TCA)

These deblocking reagents are mainly involved in DNA synthesis.


  • Infrared spectrum conforms to the standard
  • Corrosive and toxic
  • Clear liquid form

Formula: Cl₃CCOOH
MDL Number: MFCD00004177
CAS Number: 76-03-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR