Capping B 1-Methylimidazole/Tetrahydrofuran/Pyridine 1/8/1 (v/v/v), Novabiochem® for DNA/RNA synthesis
Supplier: MilliporeSigma
BI0342-0450 BI0342-4000
CABI0342-0450CS 1379 CAD
CABI0342-0450 CABI0342-4000
Capping B 1-Methylimidazole/Tetrahydrofuran/Pyridine 1/8/1 (v/v/v), Novabiochem® for DNA/RNA synthesis
Capping Reagent
These cappping reagents are involved in the formulations for oligonucleotide synthesis.
  • Soluble in water
  • Clear liquid form
  • Flammable, corrosive, and toxic effects
These are suitable for all kind of biosynthesis processes such as DNA, RNA and peptide synthesis.