Capping A THF/Pyridine/Acetic Anhydride (8/1/1), Novabiochem® for DNA/RNA synthesis
Supplier: MilliporeSigma
BI0222-0450
BI0222-4000
CABI0222-0201CS
7591.79
CAD
CABI0222-0201
CABI0222-0450
CABI0222-4000
Capping A THF/Pyridine/Acetic Anhydride (8/1/1), Novabiochem® for DNA/RNA synthesis
Capping Reagent
Capping Reagent A
- Clear liquid form
- Suitable for DNA, RNA and Oligonucleotide synthesis