Capping A THF/Lutidine/Acetic Anhydride (8/1/1), Novabiochem® for DNA/RNA synthesis
Supplier: MilliporeSigma
BI0221-0450 BI0221-2500 BI0221-4000
CABI0221-0450CS 11899.5 CAD
CABI0221-0450 CABI0221-2500 CA-BI0221-0950 CA-BI0221-4000
Capping A THF/Lutidine/Acetic Anhydride (8/1/1), Novabiochem® for DNA/RNA synthesis
Capping Reagent
Capping Reagent A
  • Clear liquid form
  • Suitable for DNA, RNA and Oligonucleotide synthesis
Acetic Anhydride: 9.9-10.1% (v/v). 3,6-Lutidine: 9.9-10.1% (v/v). Tetrahydrofuran: (79.9-80.1% (v/v).