Capping A 10% Acetic anhydride in THF, Novabiochem® for DNA/RNA synthesis
Supplier: MilliporeSigma
BI0223-0450 BI0223-4000
CABI0223-0450CS 7099.98 CAD
CABI0223-0450 CABI0223-4000
Capping A 10% Acetic anhydride in THF, Novabiochem® for DNA/RNA synthesis
Capping Reagent
Capping Reagent A for Biosynthesis
  • Clear liquid form