Water, Reagent Grade, sterile
Supplier: TEKNOVA CA
W0225
W0223
W0500
CA100216-408EA
85.13
CAD
CA100216-408
CA100220-318
CA200064-530
Water, Reagent Grade, sterile
Water
Reagent Grade Water meets the standard for ASTM TYPE with a resistance of 18 megaohms then purified by glass distillation, ion exchange, a charged carbon adsorption column and undergoes reverse osmosis, UV treatment and sterile-filtration with 0.1 micron filter.
Formula:
H₂O |
MDL Number:
MFCD00011332 CAS Number: 7732-18-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...