Diluent Solution, Sterile, Teknova
Supplier: Teknova
D5120 D5120
CA100217-524EA 317.53 CAD
CA100217-524 100217-524
Diluent Solution, Sterile, Teknova
Buffers Biological Buffers
Diluent for immunological applications.
  • 1.00% Bovine Serum Albumin
  • 0.05% (v/v) Tween-20
  • 135 mM Sodium Chloride
  • 2.7 mM Potassium Chloride
  • 4.3 mM Disodium Phosphate
  • 1.4 mM Monopotassium Phosphate