Glycerine solution 50%, sterile

Supplier: TEKNOVA CA

Synonyms: Propane-1,2,3-triolGlycerol1,2,3-Propanetriol

G1797 G1798 G1799 G1797 G1799 G1798
CA100220-042EA 247.02 CAD
CA100220-042 CA100217-622 CA100217-154 100217-154 100217-622 100220-042
Glycerine solution 50%, sterile
Glycerine

Glycerol has many uses including increasing viscosity and density of solutions.

Formula: HOCH₂CH(OH)CH₂OH
MW: 92.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00004722
CAS Number: 56-81-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR