Destain II (Methanol and Acetic Acid Solution), Teknova
Supplier: Teknova
D0020 D0020
CA101319-820EA 305.59 CAD
CA101319-820 101319-820
Destain II (Methanol and Acetic Acid Solution), Teknova
Electrophoresis Reagents Stain Removal Reagents
Polyacrylamide gel destaining solution. For use with commassie blue staining.
  • Methanol 5%, Acetic Acid 7%