Cole-Parmer® Disposable Closed XRF X-Cell Sample Cups
Supplier: SPEX
3527 3527I 3527IM 3527M 3529 3529M 3561 3561M 3565 3565M
76181-876BG 195.6 CAD
76181-876 76181-878 76181-880 76181-882 76181-884 76181-886 76181-888 76182-376 76182-378 76182-380
Cole-Parmer® Disposable Closed XRF X-Cell Sample Cups
Cuvettes Spectrophotometer Cuvettes
Fast, dependable, and reproducible XRF analysis of liquid samples.
  • One opening for fast liquid sample delivery
  • Compatible with major suppliers of XRF spectrometers
  • Contaminant-free polyethylene construction
  • Collars available to attach window film to cup for added security
Cole-Parmer® X-Cells are used throughout the world for fast, dependable, reproducible running of liquid samples in XRF spectrometers. They are designed to fit into the major suppliers of XRF spectrophotometers, such as PANalytical, Rigaku, Bruker, and Thermo Instruments. The durableMore Product Information