X-CELL® Disposable Open-Ended X-Ray Cells, 31 mm, Spex SamplePrep
Supplier: SPEX
3571 3571M
76182-270BG 195.21 CAD
76182-270 76182-272
X-CELL® Disposable Open-Ended X-Ray Cells, 31 mm, Spex SamplePrep
Cuvettes Spectrophotometer Cuvettes
Disposable open-ended X-ray cell with collar used throughout the world for fast, dependable, reproducible running of unusual and hard-to-handle samples in XRF spectrometers.
  • Ideal for unusual or hard-to-handle samples such as slurries, sludge, pastes, and viscous materials like glue, tar, and RTV sealant
  • Squared snap-ring design insures proper positioning in standard 31 mm sample holders
  • Collar holds ring in place while film is being applied
  • Can be run with open back, sealed window back, or with micro-porous film
  • Made of contaminate-free polyethylene
  • Designed to fit into the major suppliers of XRF spectrometers such as PANalytical, Rigaku, Bruker, and Thermo Instruments
Includes two polyethylene snap-rings, collar, and open body. Requires thin window film. Sold in units of 100 or 1000.