Ethidium bromide 10 mg/ml in aqueous solution

Supplier: G-Biosciences
Warning

Synonyms: 3,8-Diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide3,8-Diamino-5-ethyl-6-phenylphenanthridinium bromide

R034 R034
CA82023-294EA 397.43 CAD
82023-294 CA82023-294
Ethidium bromide 10 mg/ml in aqueous solution
Ethidium bromide
Formula: C₂₁H₂₀BrN₃
MW: 394.31 g/mol
MDL Number: MFCD00011724
CAS Number: 1239-45-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR