ABS® Undercounter Pharmacy Refrigerators, Freestanding, Premier Series, Horizon Scientific
Supplier: Horizon Scientific
PH-ABT-HC-UCFS-0204 PH-ABT-HC-UCFS-0204G PH-ABT-HC-UCFS-0504 PH-ABT-HC-UCFS-0504G
76478-940EA 2932.47 CAD
76478-940 76478-942 76478-944 76478-946
ABS® Undercounter Pharmacy Refrigerators, Freestanding, Premier Series, Horizon Scientific
Refrigerators
These Premier Series pharmacy refrigerators are freestanding and space effcient.
  • Microprocessor temperature controller
  • Digital temperature display
  • Audible and visual high and low temperature alarms
  • Remote alarm contacts
  • Keyed door lock
  • Probe access port (³/₈")
  • HFC-free refrigerant (R600a)
The Premier Series freestanding refrigerators offer a cold-storage solution to maximize storage in limited space settings. These units offer premium features for pharmacy use including a series of temperature alarms to keep stored products at maximum effectiveness,More Product Information
Order Now
SPECIFICATIONS
Model PH-ABT-HC-UCFS-0204 PH-ABT-HC-UCFS-0204G PH-ABT-HC-UCFS-0504 PH-ABT-HC-UCFS-0504G
Cat. No. 76478-940 76478-942 76478-944 76478-946
Environmentally Preferable Safer Human and Environmental Health,Energy Efficient Safer Human and Environmental Health,Energy Efficient Safer Human and Environmental Health,Energy Efficient Safer Human and Environmental Health,Energy Efficient
Display Digital Digital Digital Digital
Defrost Cycle defrost Cycle defrost Cycle defrost Cycle defrost
Access ports ³/₈" probe ³/₈" probe ³/₈" probe ³/₈" probe
Shelves/drawers 4 shelves (adjustable) 4 shelves (adjustable) 3 shelves (adjustable) 3 shelves (adjustable)
Door type 1 solid swing door, right hinged 1 glass swing door, right hinged 1 solid swing door, right hinged 1 glass swing door, right hinged
Door lock Keyed Keyed Keyed Keyed
Refrigerant HFC-free refrigerant HFC-free refrigerant HFC-free refrigerant HFC-free refrigerant
Electrical 115 V, 60 Hz, 0.90 A, 1/8 HP 115 V, 60 Hz, 0.90 A, 1/8 HP 115 V, 60 Hz, 1.3 A, 1/6 HP 115 V, 60 Hz, 1.3 A, 1/6 HP
Weight 39 kg (85 lbs.) 44 kg (97 lbs.) 59 kg (129 lbs.) 59 kg (129 lbs.)
Environmental certification Energy Star certified Energy Star certified Energy Star certified