ABS® Undercounter Pharmacy Freezers, Freestanding, Premier Series, Horizon Scientific
Supplier: Horizon Scientific
PH-ABT-HC-UCFS-0440 PH-ABT-HC-UCFS-0430 PH-ABT-HC-UCFS-0520
76478-930EA 5133.93 CAD
76478-930 76478-932 76478-934
ABS® Undercounter Pharmacy Freezers, Freestanding, Premier Series, Horizon Scientific
Freezers Laboratory / General Purpose Freezers
These Premier Series pharmacy freezers are freestanding and space effcient.
  • Microprocessor temperature controller
  • Digital temperature display
  • Audible and visual high and low temperature alarms
  • Remote alarm contacts
  • Keyed door lock
  • Probe access port (³/₈")
  • HFC-free refrigerant (R600a)
The Premier Series freestanding freezers offer a cold-storage solution to maximize storage in limited space settings. These units offer premium features for pharmacy and vaccine use including a series of temperature alarms to keep stored products at maximum effectiveness,More Product Information