ABS® Countertop Pharmacy Freezers, Freestanding, Premier Series, Horizon Scientific
Supplier: Horizon Scientific
PH-ABT-HC-UCFS-0120 PH-ABT-HC-UCFS-0120A-CAD
76478-922EA 2666.97 CAD
76478-922 76478-924
ABS® Countertop Pharmacy Freezers, Freestanding, Premier Series, Horizon Scientific
Freezers Laboratory / General Purpose Freezers
The Premier Series countertop freezers deliver quality pharmaceutical results in compact form.
  • Microprocessor temperature controller
  • Digital temperature display
  • Audible and visual high and low temperature alarms
  • Remote alarm contacts
  • Keyed door lock
  • Probe access port (³/₈")
  • HFC-free refrigerant (R600a)
The Premier Series countertop freezers offer high-quality cooling in the convenience of a compact space. These units offer premium features for pharmacy storage including a series of temperature alarms to keep stored products at maximum effectiveness.More Product Information
Order Now
SPECIFICATIONS
Model PH-ABT-HC-UCFS-0120 PH-ABT-HC-UCFS-0120A-CAD
Cat. No. 76478-922 76478-924
Environmentally Preferable Safer Human and Environmental Health,Energy Efficient Safer Human and Environmental Health,Energy Efficient
Refrigerant HFC-free refrigerant HFC-free refrigerant
Display Digital Digital
Defrost Manual Controlled Auto defrost
Electrical 115 V, 60 Hz, 1 A, 1/8 HP 115 V, 60 Hz, 1 A, 1/8 HP
Shelves/drawers 2 shelves (adjustable) 2 shelves (adjustable)
Door type 1 solid swing door, right hinged 1 solid swing door, right hinged
Door lock Keyed Keyed
Access ports ³/₈" probe ³/₈" probe
Environmental certification Energy Star certified
Weight 48 kg (106 lbs.) 48 kg (106 lbs.)