Blotting Membranes, Amersham Hybond™ ECL™, Cytiva
Supplier: Cytiva
RPN132D RPN82D
CA95038-316LPK 1439.96 CAD
CA95038-316L CA95038-426L
Blotting Membranes, Amersham Hybond™ ECL™, Cytiva
Transfer Membranes
Hybond™ ECL™ gives excellent sensitivity, resolution, and low background for all labelling and detection systems (radioactive, non radioactive, and chromogenic), especially when used with ECL™, ECL™ Prime, and ECL™ Plex Western Blotting Detection kits and reagents. Special packaging features include interleaf on both sides of the membrane and air-tight resealable aluminium bag with label space for marking the 'opened on' date.
  • 100% pure nitrocellulose membrane validated for use with all ECL™ Western blotting systems and for all protein blotting
More Product Information